Would you like to log in again?
Offset Head Aviation Snips, Proto®, ORS Nasco
Supplier: ORS Nasco
577-303LS 577-303RS
300024-488EA 47.43 USD InStock
300024-488 300024-484
Offset Head Aviation Snips, Proto®, ORS Nasco
Cutters
Snips with serrated jaw, increased control for intricate cutting.
  • Blade length of 11/4"
  • Compound leverage multiplies handle pressure
Offset handle keeps hands above working area and color coded identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty