Would you like to log in again?
Internal Pipe Wrench Sets, Proto®, ORS Nasco
Supplier: ORS Nasco
577-140SET
300023-872EA 111.26 USD InStock
300023-872
Internal Pipe Wrench Sets, Proto®, ORS Nasco
Wrenches
  • Eccentric cam action engages wrench against the walls of the pipe
  • Ideal for removing broken threaded pipe ends, plugs, and fittings
  • Installation and removal without thread damage
  • Made from hardened steel and plated to help prevent rust
  • Hex body allows for use of adjustable wrench for added torque
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty