Would you like to log in again?
Micrometer Fixed Head Torque Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-6015C 577-6013C
300027-202EA 417.79 USD InStock
300027-202 300027-200
Micrometer Fixed Head Torque Wrenches, Proto®, ORS Nasco
Wrenches
Manufactured to meet standard industrial torque applications, general industry, heavy equipment, and maintenance.
  • Calibrated to +/- 3% clockwise direction and +/- 6% counter clockwise at 20% to 100% of full scale
  • Calibration is accomplished by manual loading the torque wrenches
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty