Would you like to log in again?
Torque Multipliers, Proto®, ORS Nasco
Supplier: ORS Nasco
577-6222 577-6242 577-6252 577-6232 577-6202A 577-6234 577-6212
300027-334EA 5842.86 USD InStock
300027-334 300027-326 300027-338 300091-624 300027-340 300027-330 300027-342
Torque Multipliers, Proto®, ORS Nasco
Wrenches
Use with torque wrenches and heavy-duty power or impact sockets.
  • Standard torque accuracy is +/- 5% of reading
  • Reaction system: tubular reaction bar
  • Gear system: single stage planetary
Accessories information: Proto® torque multipliers provide overload protection with replaceable output square drives that are designed to twist off at 3%-10% over maximum output capacity
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty