Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Torque Sensing Wrenches, Utica®
Supplier: Utica®
071-TCI-250R
300017-252EA 611.74 USD InStock
300017-252
Torque Sensing Wrenches, Utica®
Wrenches
Store all adjustable models at their lowest torque setting.
  • Audible "click" and/or a few degrees of travel provide simple, fast indication of micrometer - accurate torque settings
  • Patented, low friction torque control mechanism produces accurate readings in either direction
  • Accuracy is ± 4% of setting right hand (clockwise) and ± 6% of setting left hand (counterclockwise) within upper 80% of scale
  • Two calibration adjustments (major and fine) permit easy and precise torque settings
  • Dual scale models give readings in in.-lbs./Newton Meters or ft.-lbs./Newton Meters
  • Patented spring-loaded locking collar locks scale on desired reading and remains in locked position.
  • Heavy-duty, reversible ratchet models have quick-shift lever to allow easy single-hand shifting
  • Slim, lightweight design reduces fatigue and facilitates use in confined work areas
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty