Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Torqueplus™ 12-Point Metric Combination Wrenches - Polish Finish, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1215M-T500 577-1214M-T500 577-1211M-T500 577-1207M-T500 577-1209M-T500 577-1218M-T500 577-1210M-T500 577-1224M-T500 577-1216M-T500 577-1219M-T500 577-1220M-T500 577-1222M-T500 577-1212M-T500 577-1221M-T500 577-1213M-T500 577-1217M-T500
300023-690EA 45 USD InStock
300023-690 300023-594 300023-564 300023-586 300023-720 300023-644 300023-666 300023-710 300023-656 300023-744 300023-634 300023-678 300023-624 300023-702 300023-728 300023-608
Torqueplus™ 12-Point Metric Combination Wrenches - Polish Finish, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty