Would you like to log in again?
Torqueplus™ 12-Point Short Box Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1126 577-1131 577-1122 577-1120
300023-396EA 28.21 USD InStock
300023-396 300023-412 300023-402 300023-392
Torqueplus™ 12-Point Short Box Wrenches, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Applies more torque to fastener
  • Reduces risk of fastener rounding
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty