Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Torqueplus™ 12-Point Short Combination Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1220TF 577-1222TF 577-1208TF
300023-576EA 18.96 USD InStock
300023-576 300023-732 300023-714
Torqueplus™ 12-Point Short Combination Wrenches, Proto®, ORS Nasco
Wrenches
Applies more torque to fastener.
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Reduces risk of fastener rounding
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Unique arc design increases surface contact
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty