Would you like to log in again?
Torqueplus™ 6-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1226HASD 577-1212HASD 577-1216HASD 577-1222HASD 577-1214HASD 577-1220HASD 577-1230HASD 577-1218HASD 577-1232HASD 577-1228HASD 577-1224HASD
300023-770EA 59.3 USD InStock
300023-770 300023-682 300023-762 300023-774 300023-754 300023-722 300023-658 300023-636 300023-616 300023-704 300023-738
Torqueplus™ 6-Point Combination Wrenches, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty