Would you like to log in again?
Torqueplus™ Torque Adapters, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-5120 577-5112 577-5126 577-5116 577-5122 577-5114 577-5128 577-5118 577-5124
300026-028EA 35.84 USD InStock
300026-028 300026-038 300026-042 300026-032 300026-040 300026-030 300026-036 300026-044 300026-034
Torqueplus™ Torque Adapters, Proto®, ORS Nasco
Wrenches
  • Concentric openings
  • Measures 2" from center of adapter opening to center of square drive for easy torque calculations
  • High strength, thin wall design
  • Applies more torque to fastener
  • Reduces risk of fastener rounding
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty