Would you like to log in again?
364-FHB Swivel Handle Tube Benders, Imperial Stride Tool
Supplier: Imperial Stride Tool
389-364-FHB04
300004-290EA 93.65 USD InStock
300004-290
364-FHB Swivel Handle Tube Benders, Imperial Stride Tool
Pliers
Patented series that has a swivel handle that can be repositioned midway through a bend when both handles begin to meet. Allows fabrication of bends up to 180° with reduced effort, better control and without the crossing of handles, typical of most other lever type benders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty