Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NXT® Purell® Instant Hand Sanitizer Refills, Gojo®

Supplier: GOJO
NXT® Purell®
315-2156-08 315-2156-04
300052-426CS 126.26 USD InStock
300052-426 300052-428
NXT® Purell® Instant Hand Sanitizer Refills, Gojo®
Skincare Soaps and Skin Cleansers
America's #1 instant hand sanitizer is specially formulated with moisturizers to leave hands feeling soft and refreshed.

  • Kills 99.99% of most common germs that may cause illness
  • Works in as little as 15 seconds
  • No water or towels needed
  • Alcohol-based

Weight: 2.4 lbs.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty