Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra Mild Antimicrobial Lotion Soaps w/Chloroxylenol, Gojo®

Supplier: GOJO

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

315-9212-12
300053-802CS 304.18 USD InStock
300053-802
Ultra Mild Antimicrobial Lotion Soaps w/Chloroxylenol, Gojo®
Skincare Soaps and Skin Cleansers
Lotion soap formula combines mild cleansers with 0.6% chloroxylenol (PCMX).

  • Broad-spectrum antimicrobial agent
  • For gentle handwashing and effective germ killing

Weight: 2.08 lbs.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty