Would you like to log in again?
V18™ Cordless Wet/Dry Vacuums, Milwaukee® Electric Tools
Supplier: Milwaukee® Electric Tools
V18™
495-0880-20
300041-234EA 368.78 USD InStock
300041-234
V18™ Cordless Wet/Dry Vacuums, Milwaukee® Electric Tools
Vacuum Cleaners
20% higher suction than competition.
  • Built in blower port
  • HEPA rated high efficient fiter for dust collection
  • On board hose and accessory storage
  • Quieter cordless wet/dry vacuum
  • Resuable and washable filter
  • Tool box design for excellent portability
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty