Would you like to log in again?
Utility Brush, Anchor Brand
Supplier: Anchor Brand
102-15B
300006-672EA 3.5 USD InStock
300006-672
Utility Brush, Anchor Brand
Brushes
This small hand utility brush is excellent for metal finishing in confined areas.
  • Curved wooden handle
  • Brass bristles
  • Abrasive metal brush removes corrosion, oxidation
  • Rust-resistant
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty