Would you like to log in again?
Professional Lysol® Brand Disinfectant Foam Cleaners, Reckitt Benckiser
Supplier: Reckitt Benckiser
LYSOL®
738-02775
300053-258CS 84.66 USD InStock
300053-258
Professional Lysol® Brand Disinfectant Foam Cleaners, Reckitt Benckiser
Cleaning Agents Disinfectants
  • Cleans, shines, disinfects and deodorizes
  • Contains grease-cutting agents
  • Kills staph, salmonella, pseudomonas, and HIV-1 (AIDS virus) on hard, nonporous surfaces
  • Controls growth of mold and mildew
  • Contains no CFCs
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty