Would you like to log in again?
Curved Squeegees, Magnolia Brush
Supplier: Magnolia Brush
455-4624 455-4624-TP 455-4636
300009-071EA 54.04 USD InStock
300009-071 300072-930 300009-074
Curved Squeegees, Magnolia Brush
Mops
Squeegee frame is made of 1" angle aluminum to which a 2" wide x 1/4" thick strip of high-grade black squeegee rubber or neoprene is attached with cadmium plated screws.
  • Steel bracket handle
  • Multiple handle lengths available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty