Would you like to log in again?
China Markers, Dixon® Ticonderoga
Supplier: Dixon® Ticonderoga
464-00077
300091-248PK 26.15 USD InStock
300091-248
China Markers, Dixon® Ticonderoga
Pens Pencils
  • The china marker makes beautiful bold strokes on almost any porous or non-porous surface including glass, plastic, film, paper, metal, and rubber
  • Marks wipe away clean and are paper wrapped with a tear string for easy sharpening
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty