Would you like to log in again?
Dial Calipers, General Tools
Supplier: General Tools
318-142
300008-042EA 83.37 USD InStock
300008-042
Dial Calipers, General Tools
Calipers
  • Contrasting color coded dial is graduated in .01 and 64ths with inch and mm on bar scale
  • Fractional conversion table on back
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty