Would you like to log in again?
Series 543 IDC Digimatic Indicators, Mitutoyo
Supplier: Mitutoyo
504-543-402B 504-543-472B
300082-214EA 426.9 USD InStock
300082-214 300082-216
Series 543 IDC Digimatic Indicators, Mitutoyo
Measuring and Leveling Tools
Indicator with stainless steel tip.
  • SPC output provided
  • Digital LCD
  • Measuring system inch or metric
  • Face rotates 330° for use at various angles
  • Immune to overspeed error
  • Preset, zero-set, inch/mm, +/- direction and GO/±NG functions
Stem mounting.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty