Would you like to log in again?
Stanley® Tape Rules, Stanley®
Supplier: Stanley®
680-30-496 680-30-456
300009-891EA 38.06 USD InStock
300009-891 300009-894
Stanley® Tape Rules, Stanley®
Measuring Tapes
Single sided measuring tape with polymer finish blade.
  • 26 ft./8 m length, 1" width or 16 ft./5 m length 0.75" width
  • Inch and metric
  • Hook at end
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty