Would you like to log in again?
Pouring Ladles, Extra Heavy, C.S. Osborne
Supplier: C.S. Osborne
565-373-6XH
300087-154EA 109.77 USD InStock
300087-154
Pouring Ladles, Extra Heavy, C.S. Osborne
Scoops
Ladle for extra heavy pouring.
  • Iron, lead, alumni um, even bronze and beryllium pour evenly form a forged steel bowl
  • Residue that clogs a cast-iron bowl is easily tapped out of the heat retaining steel ladle
  • Steel handles are black-smith welded to the bowls
  • Double lipped bowls
Thickness: 3/16 gauge
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty