Would you like to log in again?
2.4 V Batteries, Milwaukee® Electric Tools
Supplier: Milwaukee® Electric Tools
495-48-11-0100
300009-249EA 52.47 USD InStock
300009-249
2.4 V Batteries, Milwaukee® Electric Tools
Batteries
Milwaukee® electric tools has NiCad rechargeable dry cell (non-standard size) battery with the capacity of 1.3 Ah.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty