Would you like to log in again?
Chargers, DeWalt®
Supplier: DeWalt®
115-DC9310
300006-783EA 181.75 USD InStock
300006-783
Chargers, DeWalt®
Batteries
Dewalt® 3-stage charging system provides maximum run-time and extends overall life of the battery.
  • Diagnostics with LED indicator communicates battery charge status: charged, charging, power line problem, replace pack, and battery too hot or too cold
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty