Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
'T' Shank Metal Cutting Jig Saw Blades, DeWalt®
Supplier: DeWalt
115-DW3776-5
300060-750PK 18.55 USD InStock
300060-750
'T' Shank Metal Cutting Jig Saw Blades, DeWalt®
Razors and Blades
Unique curved tooth design minimizes stress points for longer blade life and faster cuts.
  • T-shank design fits all Bosch and DEWALT jig saws
  • Cobalt steel (bi-metal) minimize breakage
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty