Would you like to log in again?
L.S. Starrett Powerband Matrix II HSS Bi-Metal Portable Bandsaw Blades, ORS Nasco
Supplier: L.S. Starrett
681-14602
300009-948PK 48.19 USD InStock
300009-948
L.S. Starrett Powerband Matrix II HSS Bi-Metal Portable Bandsaw Blades, ORS Nasco
Razors and Blades
This blade is suitable for general purpose cutting on portable bandsaw machines.
  • Welded length and ready to use
  • Cuts harder materials
This blade fits Black & Decker, Greenlee, Milwaukee, Rockwell, Ridgid, Porter-Cable, Skil, and Unitec machine models.
Cutting edge material: HSS
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty