Would you like to log in again?
Heavy-Duty Cut-Off Wheels, Dremel®, ORS Nasco
Supplier: ORS Nasco
114-420
300006-737PK 12.93 USD InStock
300006-737
Heavy-Duty Cut-Off Wheels, Dremel®, ORS Nasco
Cutters
Heavy-duty cut-off wheels made from a hard abrasive material used for slicing and cutting metal, including hardened steel.
  • Cuts only along edge
This cutting wheels are used to cut bolts or screws, or make slots in rusted or stripped bolts and screws to allow for easy removal with a screwdriver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty