Would you like to log in again?
Bib Aprons, Anchor Brand
Supplier: Anchor Brand
101-AG-100
300076-094EA 18.7 USD InStock
300076-094
Bib Aprons, Anchor Brand
Workwear
Durable apron made from green vinyl (thickness 20 mil), ensuring long-lasting use in even the toughest environments.
  • Adjustable shoulder strap
  • Adjustable waist
  • Puncture and tear resistant
  • Waterproof
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty