Would you like to log in again?
Cut Resistant Sleeves, Memphis Glove
Supplier: Memphis Glove
127-9378T
300077-312EA 11.34 USD InStock
300077-312
Cut Resistant Sleeves, Memphis Glove
Workwear
Made of double ply 100% Kevlar.
  • High comfort level
  • Allows manual dexterity
  • Made with FDA-accepted materials for use in food processing applications
  • Launderable
  • With thumb slot
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty