Would you like to log in again?
The Shield* Safety Goggles, Jackson, Kimberly-Clark
Supplier: Kimberly-Clark
The Shield™
138-3010343
300007-063EA 47.45 USD InStock
300007-063
The Shield* Safety Goggles, Jackson, Kimberly-Clark
Goggles
Cylindrical shaped anti-fog lens made of polycarbonate offers greater range of visibility.
  • Provides extreme visual clarity in all conditions
  • Superior impact and chemical splash protection
  • Superior eye protection from 99.9% of UV radiation
  • Full face protection with detachable faceshield
  • Indirect ventilation
  • Polycarbonate curved shield conforms to face, offering increased facial protection
Weight: 0.46 lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty