Would you like to log in again?
ExoFit™ Tower Climbing Harnesses, ORS Nasco
Supplier: ORS Nasco
ExoFit™
098-1108652 098-1108651
300058-288EA 772.36 USD InStock
300058-288 300075-566
ExoFit™ Tower Climbing Harnesses, ORS Nasco
Fall Protection Fall Harnesses
It is incorporates a breathable Dri-Lex® lining that immediately draws moisture away from the body, keeping workers dry and comfortable.
  • Ergonomic design incorporates built-in shoulder, hip, and leg padding that always stays in place to enhance comfort
  • Lycra® edging moves with the wearer to prevent uncomfortable rubbing or chafing
  • The industry's first and only one-handed quick connect buckles make donning, adjusting, and removal a snap
  • Memory fit design; adjust the harness once and it stays in place
  • Wrap around, no-tangle design slips on like a vest, ensuring easy donning while providing ventilation, comfort, padding, and security
  • Sewn-in back pad and belt with side D-rings for work positioning, added comfort, and versatility
Torso buckles complete the five point adjustment system and allow for easy adjustment while securely holding the straps in place. The seat sling features cushioned padding, suspension D-rings, and a stiffener for optimal comfort and performance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty