Would you like to log in again?
Snatch Block Pulleys, DBI/Sala
Supplier: DBI/Sala
098-8003205
300058-330EA 233.79 USD InStock
300058-330
Snatch Block Pulleys, DBI/Sala
Educational Materials Physics Educational Materials Mechanics Learning Activities
Attaches to the tripod eye bolt for routing the back-up or secondary mechanical device's (winch or self retracting lifeline) cable.
Used with Tripod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty