Would you like to log in again?
ProLine™ Multi-Purpose Dry Chemical Fire Extinguishers, ABC Type, Kidde
Supplier: Kidde
ProLine™
408-466227 408-466204 408-466112-01 408-466112
300050-260EA 126.83 USD InStock
300050-260 300050-892 300050-894 300000-165
ProLine™ Multi-Purpose Dry Chemical Fire Extinguishers, ABC Type, Kidde
First Aid Supplies Fire Extinguishers
Rechargeable fire extinguishers filled with monoammonium phosphate.
  • Powder-coated cylinder for corrosion protection
  • Tough impact-resistant metal valve, handle and lever
  • Easy-to-read gauge
Fire types include common combustibles, electrical equipment, flammable liquids, gases and solvents.
Operating temperature: - 40...48.9 °C (- 40...120 °F)
Certifications: Meet most commercial and industrial specifications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty