Would you like to log in again?
AquaGuard Gravity-Flow Eye Washes, Guardian
Supplier: Guardian
333-G1540
300008-148EA 477.34 USD InStock
300008-148
AquaGuard Gravity-Flow Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Self-contained gravity flow eye wash units use gravity to deliver water through dual spray heads.
  • Top operate unit, fill unit with clean potable water and add bacteriostatic additive (supplied with unit)
  • Water flow is activated by pulling down on bright yellow eye wash tray
  • Integral full flow nozzles deliver soft spray of water, swing tray up to storage position to stop flow
Waste water is directed into drain opening in tank basin, 3 ft. long drain hose in basin directs discharge into floor drain, storage tank or other receptor. Also they are furnished with epoxy coated steel bracket for mounting unit on wall, unit can also be placed on countertop or other flat surface with green polyethylene tank material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty