Would you like to log in again?
Counter Mount Eyewash Drench Hose Units, Guardian
Supplier: Guardian
333-G5022
300068-334EA 536.23 USD InStock
300068-334
Counter Mount Eyewash Drench Hose Units, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
ANSI-compliant identification sign.
  • Swivel-type inlet
  • Valve has replaceable stainless steel seat for exceptional durability
  • Forged brass squeeze valve activated by stainless steel lever handle
Two GS-Plus™ spray heads mounted side-by-side, each head has a 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty