Would you like to log in again?
EyeSafe-X™ Faucet-Mounted Eye Washes, Guardian
Supplier: Guardian
EyeSafe-X™
333-G1101
300068-298EA 257.11 USD InStock
300068-298
EyeSafe-X™ Faucet-Mounted Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Converts any faucet into an emergency eye wash station without interfering with normal faucet operation.
  • Float off dust covers to protect outlet heads
  • Forged brass diverter valve
  • Body has ⁵⁵/₆₄"-27 female thread. furnished with 3 adaptors
  • Pull knob to activate eye wash
Water pressure holds eye wash in operation, leaving user's hands free. Outlet heads are mounted 5" apart and are adjustable aerated outlet heads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty