Would you like to log in again?
Wall Mounted Drench Hose Units, Guardian
Supplier: Guardian
333-G5025
300068-336EA 324.64 USD InStock
300068-336
Wall Mounted Drench Hose Units, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Ideal for rinsing any part of user's eye, face or body.
  • Forged brass squeeze valve activated by stainless steel lever handle
  • Valve has replaceable stainless steel seat for exceptional durability
Single FS-Plus™ spray head has 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty