Would you like to log in again?
Wall Mounted Eye Washes, Guardian
Supplier: Guardian
333-G1814
300068-314EA 461.43 USD InStock
300068-314
Wall Mounted Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Two GS-Plus™ spray-type spray heads deliver a flood of water for rinsing eyes.
  • ¹/₂" IPS chrome plated brass stay-open ball valve
Heavy duty cast aluminum wall bracket with corrosion resistant powder coated finish.
Certifications: ANSI-compliant identification sign.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty