Would you like to log in again?
High Heat Mittens, ORS Nasco
Supplier: ORS Nasco
101-K-28WL
300059-061EA 1181.76 USD InStock
300059-061
High Heat Mittens, ORS Nasco
Gloves
Lined with heat-resistant 100% wool.
  • Good abrasion resistance
  • 605 to 800 °F temperature range
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty