Would you like to log in again?
Memphis Glove® Kevlar® Gloves, with PVC Dots, ORS Nasco
Supplier: ORS Nasco
Memphis Glove®
127-9365L 127-9365S
300090-545PK 95.11 USD InStock
300090-545 300090-543
Memphis Glove® Kevlar® Gloves, with PVC Dots, ORS Nasco
Gloves
DuPont™ Kevlar® brand fiber for high tensile strength and cut resistance relative to weight, up to five times stronger than steel.
  • Will not melt, ignite, or conduct electricity
  • 18% greater cut resistance than standard Kevlar® brand fiber
  • Regular weight 7-gauge knit
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty