Would you like to log in again?
Unsupported Neoprene Gloves, Memphis Glove
Supplier: ORS Nasco
127-5430L
300061-408PK 40.16 USD InStock
300061-408
Unsupported Neoprene Gloves, Memphis Glove
Gloves
These gloves are suitable for electronic assembly, chemical labs, janitorial; biological hazards, food processing and fisheries.
  • Flock lining
  • Thick neoprene/latex blend glove
Certifications: ISO; CE approved.
Caution: Cancer and reproductive harm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty