Would you like to log in again?
No. 410 & 411 Lightweight Xenoy Safety Lockout Padlocks, Master Lock
Supplier: Master Lock
470-411RED 470-410RED
300009-177PK 106.49 USD InStock
300009-177 300009-179
No. 410 & 411 Lightweight Xenoy Safety Lockout Padlocks, Master Lock
Security Locks
Designed by safety directors specifically for OSHA lockout compliance - 1 3/4" tall.
  • Durable, lightweight, non-conductive xenoy lock body
  • Customize on-site with permanent, write-on labels
  • High security, reserved-for-safety cylinder - Up to 100,000 key changes
  • Key retaining - Ensures that padlocks are not left unlocked
  • Eight Colors Available - Add Desired Suffix - RED, YLW, BLU, GRN, BLK, ORJ, PRP, TEAL
Certifications: Compliance with OSHA " One Employee, One Lock, One Key" directive
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty