Would you like to log in again?
Safety Series™ Rotating Gate Valve Lockouts, Master Lock
Supplier: Master Lock
Safety Series™
470-480 470-481
300073-176EA 34.93 USD InStock
300073-176 300073-178
Safety Series™ Rotating Gate Valve Lockouts, Master Lock
Security Locks
Surrounds the valve operating handle, protects against accidental valve opening.
  • Unique outward rotation allows for easier installation in confined space applications
  • Each size rotates into itself to fit effectively in safety tool boxes
  • Each size nests within the next longer size to save space in storage
  • Up to 4 workers can apply their personal safety locks
  • Complete with high visibility permanent safety labels
  • Withstands chemicals; Performs effectively in extreme conditions
Temperature Range: -50.0 °F [Min], 350.0 °F [Max]
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty