Would you like to log in again?
Standard Ball Valve Lockouts, Brady
Supplier: Brady
262-65669
300065-890EA 63.8 USD InStock
300065-890
Standard Ball Valve Lockouts, Brady
Security Locks
The easy-to-use, single piece design of Ball Valve Lockouts from Brady® makes them the perfect tool for securing your valve handles in the off position.
  • Locks out ball valve sizes from 1.25 in. to 3 in. (fits standard 9/32 in. lock shackle diameters)
  • Made of powder-coated steel for superior rust resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty