Would you like to log in again?
Flammable Liquid Storage, Eagle Manufacturing
Supplier: Eagle MHC
258-1900 258-1901
300078-622EA 1406.04 USD InStock
300078-622 300078-620
Flammable Liquid Storage, Eagle Manufacturing
Furniture Storage Cabinets
Flammable liquid storage comes with adjustable galvanized shelves.
  • 2 in raised leak-proof sill
  • 2-2 in vents with baffles
  • 3-point key lock
  • Leveling legs
  • Powder coat finish
  • Corrosion resistant
Reinforced double-wall construction with 1¹/₂ in airspace.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty