Would you like to log in again?
Overpack Drums, Eagle Mfg
Supplier: Eagle Mfg
258-1600SL 258-1661 258-1601MB
300065-808EA 100.53 USD InStock
300065-808 300084-084 300065-810
Overpack Drums, Eagle Mfg
Barrels, Kegs, Drums
Designed for use with hazardous material, which is resistance against chemicals and it is made of polyethylene
  • Approved for Packing Groups I, II and III
  • Ideal for safe storage, handling and transportation of hazardous materials
  • UN Certified to HM-181
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty