Would you like to log in again?
Safety Cabinets for Combustibles
Supplier: Justrite
400-893031
300070-044EA 2180.1 USD InStock
300070-044
Safety Cabinets for Combustibles
Furniture Storage Cabinets
  • Multi-shelf cabinet provides convenient, safe storage for paints and inks
  • U-Loc™ paddle handle with slip resistant grip
  • With Sure-Grip® handle
  • 3-point self-latching, easy close doors with double key set
  • SpillSlope™ shelves safely directs spills to back and bottom into leak proof sump
  • Dual vents with flash arrestors are located on the back wall for easy connection if ventilation is needed
  • Durable, double-wall construction with 1 ¹/₂" of insulating air space
  • Durable powder coat paint
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty