Would you like to log in again?
Universal Sorbents, Anchor Brand
Supplier: Anchor Brand
103-AB-BPU100
300051-856CS 65.49 USD InStock
300051-856
Universal Sorbents, Anchor Brand
Spill Control Materials
  • Absorbs all industrial liquids (water, petroleum, and chemical based fluids) in facility
  • Dimpled & perfed
  • Perfect fit for indoor spills
  • Universal sorbents are designed for use around machinery, under leaky pipes or fittings, in messy traffic areas and for general maintenance projects
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty