Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GloWear® 8020HL Non-Certified Vests

Supplier: Ergodyne
GloWear®
150-20040
300007-172EA 5.22 USD InStock
300007-172
GloWear® 8020HL Non-Certified Vests
Workwear
Safety vests made from polyester mesh fabric (2.6 oz.).

  • Hook and loop closure
  • 1" retroreflective tape
  • Two vertical tapes
  • One size

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty