Would you like to log in again?
RS Series High Delivery Pressure Regulators, Western Enterprises
Supplier: Western Enterprises
RS Series
312-RS-11-20
300066-970EA 658.17 USD InStock
300066-970
RS Series High Delivery Pressure Regulators, Western Enterprises
Welding Supplies
  • Machined brass body and bonnet resists corrosion and extends service life
  • Dependable brass piston design
  • KEL-F™ seat assembly withstands high service pressures
  • Dual inlet filtering extends regulator life
  • Outlet 180° opposed to inlet
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty